Merck
Bioactive iridoids and a new lignan from Allamanda cathartica and Himatanthus fallax from the Suriname rainforest.
Journal of natural products (1998-01-15)
M S Abdel-Kader, J Wisse, R Evans, H van der Werff, D G Kingston
RÉSUMÉ
Bioassay-guided fractionation of the EtOAc extract of both Allamanda cathartica and Himatanthus fallax (Apocynaceae) using the Sc-7 yeast strain resulted in the isolation of the weakly cytotoxic isoplumericin and plumericin. In addition, the new lignan 7(R)-methoxy-8-epi-matairesional and three known compounds, plumieride, matairesinol, and pinoresinol, were isolated from H. fallax.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Plumieride, ≥95% (LC/MS-ELSD)