- Hepatocellular carcinoma and focal nodular hyperplasia associated with norethandrolone-therapy: a case report.
Hepatocellular carcinoma and focal nodular hyperplasia associated with norethandrolone-therapy: a case report.
Blut (1986-02-01)
S Serke, D Dienemann, B Speck, R Zimmermann, U Baer, D Huhn
PMID3004620
RÉSUMÉ
A 33-year-old woman was treated for severe aplastic anemia with norethandrolone over a period of four years, with a cumulative dose of 25 g. In the fifth year of therapy two intrahepatic tumors were detected and were classified as hepatocellular carcinoma and as focal nodular hyperplasia, respectively.