Merck
  • Efficient targeting and tumor retardation effect of pancreatic adenocarcinoma up-regulated factor (PAUF)-specific RNA replacement in pancreatic cancer mouse model.
Efficient targeting and tumor retardation effect of pancreatic adenocarcinoma up-regulated factor (PAUF)-specific RNA replacement in pancreatic cancer mouse model.
Cancer letters (2013-11-06)
Yun-Hee Kim, Ju Young Moon, Eun-Ok Kim, Sang-Jin Lee, Se Hun Kang, Seok Ki Kim, Kyun Heo, Yusun Lee, Hana Kim, Kyung-Tae Kim, Daehong Kim, Min Sun Song, Seoung-Wook Lee, Yangsoon Lee, Sang Seok Koh, In-Hoo Kim
RÉSUMÉ
The soluble protein pancreatic adenocarcinoma up-regulated factor (PAUF) plays an important role in pancreatic tumor progression and has begun to attract attention as a therapeutic target for pancreatic cancer. We herein present PAUF RNA-targeting gene therapy strategies with both targeting and therapeutic function using trans-splicing ribozyme (TSR) in pancreatic cancer. We developed adenoviral PAUF-targeting TSR (Rz) containing a PAUF-specific internal guide sequence (IGS) determined by library screening. This Rz harbors suicide gene, herpes simplex virus thymidine kinase (HSV-tk) or firefly luciferase (Luc) as a transgene for 3' exon replacement of PAUF RNAs. Ad-Rz-TK, Rz harboring the HSV-tk, showed significant inhibition of tumor growth in vivo as well as PAUF-dependent cell death in vitro via a successful trans-splicing reaction. Selective induction of Rz-controlled transgene in PAUF-expressing pancreatic cancer was confirmed through noninvasive in vivo imaging; a luminescence signal from Rz harboring Luc (Ad-Rz-Luc) was detectable only in pancreatic tumor sites, not in normal mice. In addition, a [(125)I] FIAU signal reflecting thymidine kinase expression through SPECT and ex vivo biodistribution was co-localized with the tumor sites when we treated with Ad-Rz-TK in orthotopic xenograft model. Taken together, these results imply that PAUF-targeting TSR can contribute to successful targeted gene therapy for pancreatic cancer.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Fialuridine, ≥98% (HPLC)