Merck
Relationship between 3-O-methyldopa and the clinical effects of entacapone in advanced Parkinson's disease.
Hiroshima journal of medical sciences (2011-11-08)
Aiko Ishihara, Takafumi Miyachi, Takeshi Nakamura, Toshiho Ohtsuki, Yasuhiro Kimura, Kenji Kihira, Takemori Yamawaki, Masayasu Matsumoto
RÉSUMÉ
The aim of this study is to clarify the relationship between serum 3-O-methyldopa (3-OMD) and the clinical effects of entacapone. The 3-OMD and maximum serum concentration (Cmax) of levodopa were measured in 21 Parkinson's Disease patients who took 100 mg levodopa / dopa decarboxylase inhibitor. After the administration of entacapone, the 3-OMD concentration and percentage of "on" time during waking hours (% of "on" time) were studied for 8 weeks. The 3-OMD concentration was reduced by 34%, and the increase in % of "on" time was 28% at the 8th week compared with baseline. We defined the COMT-index as [baseline 3-OMD concentration] / [levodopa Cmax when 100 mg levodopa was administered alone]. The COMT-index was significantly correlated with the increase in % of "on" time at the 8th week. In conclusion, the measurement of baseline 3-OMD and levodopa pharmacokinetics is useful for predicting the clinical effects of entacapone.
MATÉRIAUX
Référence du produit
Marque
Description du produit
USP
Levodopa Related Compound B, United States Pharmacopeia (USP) Reference Standard