Merck
A feruloyltyramine trimer isolated from potato common scab lesions.
Phytochemistry (2010-10-30)
Russell R King, Larry A Calhoun
RÉSUMÉ
(1)H NMR analysis established that a potential suberin intermediate isolated from potato common scab lesions contained three O-methyl groups, a phenylcoumaran-type linkage and a conjugated trans double bond. Mass spectral data determined its molecular formula as indicative of a dehydrotrimer structure formed from three feruloyltyramine units. (1)H and (13)C NMR correlation studies supported the structure as that of a grossamide unit (3) linked through its double bond to the feruloyl phenolic of a third feruloyltyramine group. Identification of the feruloyltyramine trimer (4) expands the number of cross-linked intermediates potentially involved in the suberization process and highlights the presence of a second type of inter-unit linkage available for synthesis of the poly-phenolic domains.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Moupinamide, ≥95% (LC/MS-ELSD)