Merck
Determination of 6-oxo-morphinans, as the oximes, by difference circular dichroism spectroscopy.
Analytical and bioanalytical chemistry (2002-10-10)
A Szentesi, A Gergely, P Horváth, S Hosztafi, Gy Szász
RÉSUMÉ
A negative Cotton effect is observed in the circular dichroism (CD) spectra of 6-oxo-morphinans in the wavelength range of n-pi* electron transitions. 6-oxo-Morphinans can be transformed into oxime derivatives with hydroxylamine and after oxime formation the CD spectra are significantly different. Oxime formation was monitored by CD and by HPLC. It was established that under the experimental conditions used oxime formation was complete within 90 min. The method suggested for the determination of 6-oxo-morphinans is based on the considerable differences between the ellipticities before and after the oxime formation. The ellipticity difference varies linearly with concentration in the range 2 x 10(-5)-5 x 10(-4) mol L(-1) for the three 6-oxo-morphinans examined (oxycodone, hydrocodone, and 14-hydroxycodeinone). For hydrocodone the dependence is also linear in a lower concentration range (5 x 10(-6)-10(-4) mol L(-1)). The new difference CD spectroscopic method can be applied to the selective determination of 6-oxo-morphinans in bulk and dosage forms.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Oxycodone impurity D, European Pharmacopoeia (EP) Reference Standard
Le tarif et la disponibilité ne sont pas disponibles actuellement.