Merck
Au(III)/TPPMS-catalyzed benzylation of indoles with benzylic alcohols in water.
The Journal of organic chemistry (2013-11-22)
Hidemasa Hikawa, Hideharu Suzuki, Isao Azumaya
RÉSUMÉ
A novel and efficient method for the Au(III)/TPPMS-catalyzed direct substitution reaction of benzhydryl and benzylic alcohols with indoles in water is developed. Au(III)/TPPMS is an effective catalyst for the benzylation of the strong π nucleophile 1-methylindole, while common Brønsted or Lewis acids are ineffective.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Methylindole, ≥97%