Merck
Synthesis and biological evaluation of DL-4,4-difluoroglutamic acid and DL-gamma,gamma-difluoromethotrexate.
Journal of medicinal chemistry (1996-01-05)
T Tsukamoto, T Kitazume, J J McGuire, J K Coward
RÉSUMÉ
DL-4,4-Difluoroglutamic acid (DL-4,4-F2Glu) and its methotrexate analogue, DL-gamma,gamma-difluoromethotrexate (DL-gamma,gamma-F2MTX), were synthesized and evaluated as alternate substrates or inhibitors of folate-dependent enzymes. Synthesis of DL-4,4-F2Glu involved the nitroaldol reaction of ethyl nitroacetate with a difluorinated aldehyde ethyl hemiacetal as a key step. Attempted ligation of DL-4,4-F2Glu to methotrexate (MTX), catalyzed by human folylpoly-gamma-glutamate synthetase (FPGS), revealed that DL-4,4-F2Glu is a poor alternate substrate. DL-gamma,gamma-F2MTX was synthesized by a route proceeding through N-[4-(methylamino)benzoyl]-4,4-difluoroglutamic acid di-tert-butyl ester followed by alkylation with 6-(bromomethyl)-2,4-pteridinediamine hydrobromide. DL-gamma,gamma-F2MTX was found to be neither a substrate nor an inhibitor of human FPGS. The fluorinated analogue of MTX, however, inhibits DHFR and cell growth with the same potency as MTX.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Ethyl nitroacetate, 97%