Merck
Stability and toxicity of selected chlorinated benzophenone-type UV filters in waters.
Acta chimica Slovenica (2013-12-24)
Rensheng Zhuang, Romina Žabar, Gorica Grbović, Darko Dolenc, Jun Yao, Tatjana Tišler, Polonca Trebše
RÉSUMÉ
In our study, the transformation of two most widely used UV filters, benzophenone-3 (BP3) and benzophenone-4 (BP4), in chlorinated water with disinfection reagents sodium hypochlorite (NaClO) and trichloroisocyanuric acid (TCCA) was studied. Based on the HPLC/MS and UV-Vis analysis the formation of two different chlorinated products (5-chloro-2-hydroxy-4-methoxybenzophenone and 3,5-dichloro-2-hydroxy-4-methoxybenzophenone) was established. Identity of chlorinated products was confirmed by means of comparison of retention times with independently synthesized standards. Photostability study showed that dichloro-derivative in water is less stable then parent compounds, which is not the case for monochloro-derivatives. Toxicity of chlorinated compounds tested by Vibrio fischeri was found to be in the same range as that of the starting compounds. Preliminary testing of real water samples from swimming pools and sea swimming areas confirmed the presence of BP3 and its 3,5-dichloro derivative.