Merck
  • Spectroscopic probing of the effect of alkanols on the properties of the head group region in reverse micelles of AOT-heptane-water.
Spectroscopic probing of the effect of alkanols on the properties of the head group region in reverse micelles of AOT-heptane-water.
Journal of colloid and interface science (2003-03-06)
Elsa Abuin, Eduardo Lissi, Angela Cerón, María Angélica Rubio
RÉSUMÉ
The effects of addition of alkanols (ethanol, n-hexanol, and 3-ethyl-3-pentanol) on the micropolarity and microviscosity of the head group region in reverse micelles of AOT-heptane-water have been investigated by fluorescence probing methods (ANS fluorescence yield and TMADPH fluorescence anisotropy), complemented by the use of the solvatochromic probe E(T)(30) in absorption spectroscopy. For all the alkanols considered, ANS fluorescence in AOT reverse micelles (at W=3) is quenched by additive incorporation, being the effect elicited almost independent of the alkanol chain length and topology. As sensed by the E(T)(30) parameter, the micropolarity of the micelle surface increases, remains unmodified, and decreases upon addition of ethanol, 3-ethyl-3-pentanol, and hexanol, respectively. While ethanol barely modifies the fluorescence anisotropy of TMADPH, 3-ethyl-3-pentanol and n-hexanol addition strongly decrease it. The similarity of the tendencies of ANS data to TMADPH anisotropies and the differences between ANS data and E(T)(30) values would indicate that, at least for 3-ethyl-3-pentanol and n-hexanol, microviscosity, rather than micropolarity, must be considered to interpret the effect of the alkanols upon the fluorescent behavior of ANS.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-Ethyl-3-pentanol, 98%