Merck
Influence of DNA repair deficiencies on the UV sensitivity of yeast cells in different cell cycle stages.
Mutation research (1990-12-01)
W Siede, E C Friedberg
RÉSUMÉ
Synchronously dividing haploid yeast cells were UV-irradiated in various stages of the cell cycle after release from alpha-factor arrest. In confirmation of earlier results (Chanet et al., 1973), in wild-type strains G1/S phase cells were found to be the most sensitive and late S/G2 cells the most resistant. Stationary-phase (G0) cells were significantly more UV resistant than G1 cells. Strains defective in nucleotide excision repair lost enhanced resistance in the G2 phase and were most UV-sensitive in the G0 state. Reduced G2 resistance was also observed in rad6 mutants but not in rad9 mutants. After UV-irradiation in G1 phase rad9 mutant cells showed a reduced G1/S phase arrest.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Trp-His-Trp-Leu-Gln-Leu, ≥97% (HPLC)