Merck
  • Accurate quantification of dimethylamine (DMA) in human urine by gas chromatography-mass spectrometry as pentafluorobenzamide derivative: evaluation of the relationship between DMA and its precursor asymmetric dimethylarginine (ADMA) in health and disease.
Accurate quantification of dimethylamine (DMA) in human urine by gas chromatography-mass spectrometry as pentafluorobenzamide derivative: evaluation of the relationship between DMA and its precursor asymmetric dimethylarginine (ADMA) in health and disease.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2006-10-03)
Dimitrios Tsikas, Thomas Thum, Thomas Becker, Vu Vi Pham, Kristina Chobanyan, Anja Mitschke, Bibiana Beckmann, Frank-Mathias Gutzki, Johann Bauersachs, Dirk O Stichtenoth
RÉSUMÉ
Dimethylamine [DMA, (CH(3))(2)NH)] is abundantly present in human urine. Main sources of urinary DMA have been reported to include trimethylamine N-oxide, a common food component, and asymmetric dimethylarginine (ADMA), an endogenous inhibitor of nitric oxide (NO) synthesis. ADMA is excreted in the urine in part unmetabolized and in part after hydrolysis to DMA by dimethylarginine dimethylaminohydrolase (DDAH). Here we describe a GC-MS method for the accurate and rapid quantification of DMA in human urine. The method involves use of (CD(3))(2)NH as internal standard, simultaneous derivatization with pentafluorobenzoyl chloride and extraction in toluene, and selected-ion monitoring of m/z 239 for DMA and m/z 245 for (CD(3))(2)NH in the electron ionization mode. GC-MS analysis of urine samples from 10 healthy volunteers revealed a DMA concentration of 264+/-173 microM equivalent to 10.1+/-1.64 micromol/mmol creatinine. GC-tandem MS analysis of the same urine samples revealed an ADMA concentration of 27.3+/-15.3 microM corresponding to 1.35+/-1.2 micromol/mmol creatinine. In these volunteers, a positive correlation (R=0.83919, P=0.0024) was found between urinary DMA and ADMA, with the DMA/ADMA molar ratio being 10.8+/-6.2. Elevated excretion rates of DMA (52.9+/-18.5 micromol/mmol creatinine) and ADMA (3.85+/-1.65 micromol/mmol creatinine) were found by the method in 49 patients suffering from coronary artery disease, with the DMA/ADMA molar ratio also being elevated (16.8+/-12.8). In 12 patients suffering from end-stage liver disease, excretion rates of DMA (47.8+/-19.7 micromol/mmol creatinine) and ADMA (5.6+/-1.5 micromol/mmol creatinine) were found to be elevated, with the DMA/ADMA molar ratio (9.17+/-4.2) being insignificantly lower (P=0.46). Between urinary DMA and ADMA there was a positive correlation (R=0.6655, P<0.0001) in coronary artery disease, but no correlation (R=0.27339) was found in end-stage liver disease.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2,3,4,5,6-Pentafluorobenzoyl chloride, 99%