- Improved synthesis of meso-aryl-substituted [26]hexaphyrins.
Improved synthesis of meso-aryl-substituted [26]hexaphyrins.
Organic letters (2003-10-11)
Masaaki Suzuki, Atsuhiro Osuka
PMID14535749
RÉSUMÉ
[reaction: see text] Acid-catalyzed reactions of 5-(2',3',4',5',6'-pentafluorophenyl)dipyrromethane with aryl aldehydes and of 5-(aryl)dipyrromethanes with 2,3,4,5,6-pentafluorobenzaldehyde allowed the preparation of a variety of meso-aryl [26]hexaphyrins.