Merck
Decreasing profile of residual sulphaquinoxaline in eggs.
British poultry science (1998-07-03)
N Furusawa, Y Tsuzukida, H Yamaguchi
RÉSUMÉ
1. Sulphaquinoxaline (SQ) was added to the diet of laying hens at 200 mg/kg for 7 successive days. Contents (mg/kg) of SQ in albumen and egg yolk of eggs laid after drug withdrawal were determined by high pressure liquid chromatography (HPLC). The contents in the whole egg were calculated taking into consideration the respective weights of albumen and egg yolk. 2. A time-lag in the initiation of decrease of SQ from eggs after the withdrawal of dietary SQ was observed. 3. The decreasing pattern from whole egg could be well described by the following equation with a time-lag of 1.0 d, y = 2.07 e-0.5620(t-1.0), where y is the SQ content in whole egg, t is time (d) after the withdrawal of dietary SQ and a constant of 2.07 is the SQ content in whole egg laid at the withdrawal. 4. Biological half-life of SQ in the whole egg was estimated to be 1.23 d. 5. From the above equation, SQ residue of whole egg laid at 9th d after withdrawal will be below the detection limit of 0.01 mg/kg.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Sulfaquinoxaline, PESTANAL®, analytical standard