Merck
Purification and partial characterization of the OmpA family of proteins of Pasteurella haemolytica.
Infection and immunity (1997-01-01)
P J Mahasreshti, G L Murphy, J H Wyckoff, S Farmer, R E Hancock, A W Confer
RÉSUMÉ
This study was conducted to partially characterize and identify the purity of two major outer membrane proteins (OMPs) (with molecular weights of 32,000 and 35,000 [32K and 35K, respectively]) of Pasteurella haemolytica. The 35K and 32K major OMPs, designated Pasteurella outer membrane proteins A and B (PomA and PomB, respectively), were extracted from P. haemolytica by solubilization in N-octyl polyoxyl ethylene. The P. haemolytica strain used was a mutant serotype A1 from which the genes expressing the 30-kDa lipoproteins had been deleted. PomA and PomB were separated and partially purified by anion-exchange chromatography. PomA but not PomB was heat modifiable. The N-terminal amino acid sequences of the two proteins were determined and compared with reported sequences of other known proteins. PomA had significant N-terminal sequence homology with the OmpA protein of Escherichia coli and related proteins from other gram-negative bacteria. Moreover, polyclonal antiserum raised against the E. coli OmpA protein reacted with this protein. PomA was surface exposed, was conserved among P. haemolytica biotype A serotypes, and had porin activity in planar bilayers. No homology between the N-terminal amino acid sequence of PomB and those of other known bacterial proteins was found. Cattle vaccinated with live P. haemolytica developed a significant increase in serum antibodies to partially purified PomA, as shown by enzyme-linked immunosorbent assays, and to purified PomA and PomB, as detected on Western blots and by densitometry.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Poly(ethylene glycol) octyl ether