Merck
  • Agonist-induced functional desensitization of the mu-opioid receptor is mediated by loss of membrane receptors rather than uncoupling from G protein.
Agonist-induced functional desensitization of the mu-opioid receptor is mediated by loss of membrane receptors rather than uncoupling from G protein.
Molecular pharmacology (1996-11-01)
Y Pak, A Kouvelas, M A Scheideler, J Rasmussen, B F O'Dowd, S R George
RÉSUMÉ
The effects of acute exposure of the opioid peptide [D-Ala2,N-MePhe4, Gly-ol5]enkephalin (DAMGO) on the mu-opioid receptor were examined in Chinese hamster ovary (CHO) K-1 and baby hamster kidney stable transfectants. In the CHO cell line, acute 1-hr treatment with DAMGO decreased the density of receptors without affecting the affinity or proportion of agonist-detected sites and attenuated the ability of the agonist to inhibit forskolin-stimulated cAMP accumulation. In contrast, similar 1-hr treatment of baby hamster kidney cells did not affect receptor density or agonist ability to inhibit cAMP accumulation, but longer duration of agonist exposure resulted in a reduction in membrane receptor, identical to the CHO cells. These results suggested that for the mu-opioid receptor, alteration in receptor density was the major determinant for the observed agonist-induced desensitization. Consistent with this notion, the ratio of the DAMGO concentration yielding half-maximal occupation of the mu receptor to that yielding half-maximal functional response was < 1. This suggests the necessity for a high mu receptor occupancy rate for maximal functional response, so that any loss of cell surface opioid-binding sites was a critical determinant in reducing the maximal response. This hypothesis was further supported by the observation that irreversible inactivation of fixed proportions of opioid-binding sites with beta-chlorn-altrexamine demonstrated that there were few spare receptors, which is in contrast to what has been reported for other G protein-coupled receptors, including the delta-opioid receptor. Taken together, these data suggest that the opioid agonist DAMGO has a high affinity for the mu receptor but must occupy > 70% of the available receptors to generate the maximal second messenger-linked response.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
β-Chlornaltrexamine dihydrochloride, solid