Merck
Determination of cobalt in vegetable animal foodstuffs by differential pulse adsorptive voltammetry using alpha-benzil dioxime.
The Science of the total environment (1995-12-22)
S Giroussi, A Voulgaropoulos, A K Ayiannidis, J Golimowski, M Janicki
RÉSUMÉ
A selective and sensitive voltammetric method for the determination of cobalt in vegetable animal foodstuffs is developed. The method is based on the use of alpha-benzil dioxime (alpha-BD) as a chelating agent for differential pulse adsorptive stripping voltammetry (DPASV) and is free from zinc interferences. The influence of pH, time and alpha-BD concentration on the peak resolution and height are discussed. The method was successfully applied in some typical vegetable animal foodstuffs with R.S.D. < 6%.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
anti-Diphenylglyoxime, 97%