Merck
Norpethidine toxicity and patient controlled analgesia.
British journal of anaesthesia (1993-11-01)
P A Stone, P E Macintyre, D A Jarvis
RÉSUMÉ
Patient-controlled analgesia (PCA) with i.v. opioids is prescribed increasingly. We report three cases of norpethidine toxicity in patients receiving pethidine by PCA.