Merck
Neoadjuvant (induction) chemotherapy in advanced bladder carcinoma.
International urology and nephrology (1993-01-01)
L Türkeri, Y Ilker, F Simşek, A Akdaş
RÉSUMÉ
Traditional approaches for the treatment of locally advanced bladder tumours may not be sufficient enough. Neoadjuvant chemotherapy, a new modality, may be beneficial by enhancing the local and systemic control of the disease. The background of this new modality and early results of various trials are discussed.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
cis-Dichlorobis(dimethyl sulfoxide)platinum(II), 97%