- Application of N,N-dimethylformamide dineopentyl acetal for efficient anchoring of N alpha-9-fluorenylmethyloxycarbonylamino acids as p-alkoxybenzyl esters in solid-phase peptide synthesis.
Application of N,N-dimethylformamide dineopentyl acetal for efficient anchoring of N alpha-9-fluorenylmethyloxycarbonylamino acids as p-alkoxybenzyl esters in solid-phase peptide synthesis.
International journal of peptide and protein research (1984-04-01)
F Albericio, G Barany
PMID6547115
RÉSUMÉ
The attachment of Fmoc-amino acids onto p- alkoxybenzyl alcohol resins via DCC-DMAP coupling suffers from two different problems: formation of dimers and racemization. The use of N,N-dimethylformamide dineopentyl acetal for the preparation of Fmoc- aminoacyloxybenzyl handles is the basis of a safe and efficient anchoring method that avoids both problems.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Le tarif et la disponibilité ne sont pas disponibles actuellement.