- Effect of anionic polymeric hydrogels on spermatozoa motility.
Effect of anionic polymeric hydrogels on spermatozoa motility.
Biomaterials (1984-09-01)
H Singh, M S Jabbal, A R Ray, P Vasudevan
PMID6487714
RÉSUMÉ
The effects of a few synthetic polymers on the motility of human spermatozoa in vitro have been studied. An alternate copolymer of styrene and maleic anhydride, poly(S-MA), poly (styrene-maleic acid), poly(S-MC), poly(hydroxy-ethyl methacrylate-methacrylic acid) copolymer, poly(HEMA-MAC), poly(HEMA) homopolymer and poly(MAC) homopolymer were chosen for this purpose. It was found that all the carboxylic acid containing polymers are strong inhibitors of the motility of spermatozoa. Poly(HEMA) did not have any inhibitory effect on the motility of spermatozoa.
MATÉRIAUX
Numéro du produit