- [Synthesis of the leukotriene A4 methyl ester via acetylene intermediates].
[Synthesis of the leukotriene A4 methyl ester via acetylene intermediates].
Bioorganicheskaia khimiia (1987-08-01)
Iu Iu Belosludtsev, G I Miagkova, R P Evstigneeva, N I Bobrova, K K Pivnitskiĭ
PMID2823834
RÉSUMÉ
The strategy of acyclic eicosanoid synthesis via polyacetylenic intermediates is examplified by the synthesis of the racemic leukotriene A4 methyl ester. Leukotriene synthons, namely, trideca-1,4,7-triyne and methyl 6-formyl-5,6-trans-epoxyhexanoate, were synthesised using propargylic alcohol (thrice) and 1-heptyne as starting materials. In the course of the synthesis all new carbon-carbon bonds were created through acetylenide anion condensations and (Z)-double bonds are introduced by triple bond hydrogenations. The strategy provides a straightforward and stereospecific synthetic pathway.