Merck
Effects of intraduodenal injection of Lactobacillus brevis SBC8803 on autonomic neurotransmission and appetite in rodents.
Neuroscience letters (2013-02-09)
Yuko Horii, Yasukazu Nakakita, Yoshiyuki Fujisaki, Saori Yamamoto, Nanako Itoh, Koyomi Miyazaki, Hirotaka Kaneda, Katsutaka Oishi, Tatsuro Shigyo, Katsuya Nagai
RÉSUMÉ
Lactobacilli provide several health benefits to mammals, including humans. We previously observed that in rats, intraduodenal injection of Lactobacillus johnsonii La1 elevated efferent gastric vagal nerve activity (efferent-GVNA), while Lactobacillus paracasei ST11 suppressed efferent-GVNA, and thereby increased or decreased food intake. To determine the function of Lactobacillus brevis (SBC8803), its effect on food intake was examined by providing food containing heat-killed SBC8803 to mice. We observed that administration of SBC8803 elevated food intake. Because the afferent intestinal vagal nerve (IVN) is hypothesized to be involved in efferent-GVNA changes, we examined the effect of intraduodenal administration of heat-killed SBC8803 on efferent-GVNA and afferent-IVN activity (IVNA) in rats. In this study, we found that intraduodenal administration of heat-killed SBC8803 increased both efferent-GVNA and afferent-IVNA in rats. Moreover, IV administration of the serotonin 3 receptor antagonist granisetron eliminated the effects of SBC8803 on efferent-GVNA and afferent-IVNA. These findings suggest that heat-killed SBC8803 enhances appetite by elevating digestion and absorption abilities via changes in autonomic neurotransmission that might be mediated by the serotonin 3 receptor.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Granisetron hydrochloride, ≥98% (HPLC), solid
Granisetron hydrochloride, European Pharmacopoeia (EP) Reference Standard