Merck
Investigation of inclusion complex of cilnidipine with hydroxypropyl-β-cyclodextrin.
Carbohydrate polymers (2012-09-05)
Liandong Hu, Hailei Zhang, Weihua Song, Deliang Gu, Qiaofeng Hu
RÉSUMÉ
The objective of this study was to improve the water-solubility and photostability of cilnidipine by complexing it with hydroxypropyl-β-cyclodextrin (HP-β-CD or HP-beta-CD). The interactions of cilnidipine and HP-β-CD were characterized by ultra violet-visible (UV/VIS) spectroscopy, differential scanning calorimetry (DSC), powder X-ray diffraction (PXRD), Fourier transformation-infrared (FT-IR) spectroscopy and (1)H nuclear magnetic resonance ((1)H NMR) spectroscopy to verify the formation of cilnidipine-HP-β-CD complex inclusion. Moreover, the binding sites in the HP-β-CD structure were also tracked through (1)H NMR spectroscopy analysis. All the characterization information proved the formation of cilnidipine-HP-β-CD inclusion complex, and the results demonstrated the superiority of the inclusion complex in dissolution rates and photostability; in addition, the apparent solubility of cilnidipine was increased more than 10,000-fold in the presence of HP-β-CD. The stability constant (1:1) was found to be 50,116 M(-1), suggesting a high tendency of the drug to enter the HP-β-CD cavity. These results identified the cilnidipine-HP-β-CD inclusion complex as an effective new approach to design a novel formulation for pharmaceutical application.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Cilnidipine, ≥98% (HPLC), powder