Merck
Design and synthesis of biotinylated inositol 1,3,4,5-tetrakisphosphate targeting Grp1 pleckstrin homology domain.
Bioorganic & medicinal chemistry (2011-10-15)
Kensaku Anraku, Teruhiko Inoue, Kenji Sugimoto, Kota Kudo, Yoshinari Okamoto, Takashi Morii, Yasuo Mori, Masami Otsuka
RÉSUMÉ
A bifunctional molecule containing biotin and d-myo-inositol 1,3,4,5-tetrakisphosphate was synthesized. This molecule was designed on the basis of X-ray structure of the complex of d-myo-inositol 1,3,4,5-tetrakisphosphates, Ins(1,3,4,5)P(4), and Grp1 PH (general receptor of phosphoinositides pleckstrin homology) domain for the application to the widely employed biotin-avidin techniques. The building block of inositol moiety was synthesized starting with myo-inositol and assembled with the biotin-linker moiety through a phosphate linkage. The equilibrium dissociation constant K(D) of biotinylated Ins(1,3,4,5)P(4) binding of original Grp1 PH domain was 0.14 μM in pull-down analysis, which was comparable to that of unmodified Ins(1,3,4,5)P(4). Furthermore, biotinylated Ins(1,3,4,5)P(4) had an ability to distinguish Grp1 PH domain from PLCδ(1) PH domain. Thus, biotinylated Ins(1,3,4,5)P(4) retained the binding affinity and selectivity of original Grp1 PH domain, and realized the intracellular Ins(1,3,4,5)P(4) despite a tethering at the 1-phosphate group of inositol.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
D-myo-Inositol 1,3,4,5-tetrakis(phosphate) ammonium salt, ≥95%