Merck
[Determination of nequinate and buquinolate in livestock products using liquid chromatography-tandem mass spectrometry].
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2011-07-02)
Takayuki Nakajima, Hiroshi Hayashi, Takeo Sasamoto, Maki Kanda, Tomoko Kusano, Youko Matsushima, Setsuko Kanai, Kazue Takeba, Toshihiro Nagayama
RÉSUMÉ
We studied the simultaneous determination of nequinate and buquinolate, which are used as feed additives to prevent coccidiosis, by means of liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS). The sample was extracted with acetonitrile, then loaded onto an HLB mini-column with 20% methanol. After clean-up with 20% methanol, the analytes were eluted with acetonitrile-methanol (1 : 1). The coccidiostats in the purified samples were determined using ESI-MRM mode LC-MS/MS with a sample matrix calibration curve. Mean recoveries of nequinate and buquinolate from 8 kinds of livestocks samples (chicken muscle, chicken liver, chicken heart, swine muscle, swine heart, cattle muscle, sheep muscle, egg) were in the range of 89.5% to 108.6%, and the relative standard deviation values were <20% (n=10) at the levels of 0.01 μg/g and 0.05 μg/g, respectively. The limits of quantification of these compounds were 0.001 μg/g in each sample.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Supelco
Nequinate, VETRANAL®, analytical standard
Supelco
Buquinolate, VETRANAL®, analytical standard