Merck
Novel one-pot three component reaction for the synthesis of [2-(alkylsulfanyl)imidazo[1,2-a]pyridin-3-yl](aryl)methanone.
Journal of combinatorial chemistry (2009-11-13)
Ebrahim Kianmehr, Mohammad Ghanbari, Mehri Nadiri Niri, Reza Faramarzi
RÉSUMÉ
A one-pot, three-component reaction between pyridine, phenacyl bromide, and thiocyanate is described. The reaction afforded the corresponding special type of fully substituted imidazo[1,2-a]pyridine derivatives in good yields without using any catalyst or activation.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Imidazo[1,2-a]pyridine, 99%