Merck
The use of intravenous aminobisphosphonates for the treatment of Paget's disease of bone.
Mini reviews in medicinal chemistry (2009-08-20)
Luigi Gennari, Daniela Merlotti, Giuseppe Mossetti, Domenico Rendina, Vincenzo De Paola, Giuseppe Martini, Ranuccio Nuti
RÉSUMÉ
Paget's disease of bone is a focal skeletal disorder characterized by the formation of structurally abnormal bone, deformity and other complications leading to significant disability and bone pain. Recently, the availability of newer, more potent nitrogen-containing bisphosphonates has improved treatment outcomes, allowing a more effective and convenient management of this disorder.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Neridronate, ≥97% (NMR), solid