Merck
  • Metabolism of two new benzodiazepine-type anti-leishmanial agents in rat hepatocytes and hepatic microsomes and their interaction with glutathione in macrophages.
Metabolism of two new benzodiazepine-type anti-leishmanial agents in rat hepatocytes and hepatic microsomes and their interaction with glutathione in macrophages.
The Journal of pharmacy and pharmacology (2009-02-19)
M Dao Duong Thi, M Helen Grant, Alexander B Mullen, Justice N A Tettey, Simon P MacKay, Rachel L Clark
RÉSUMÉ
To measure the metabolism and toxicity of 7-chloro-4-(cyclohexylmethyl)-1-methyl-3,4-dihydro-1H-1,4-benzodiazepine-2,5-dione (BNZ-1) and 4-cyclohexylmethyl-1-methyl-3,4-dihydro-1H-1,4-benzodiazepine-2,5-dione (BNZ-2), two new benzodiazepine analogues found to be effective against Leishmania amastigotes in vitro. The metabolism of BNZ-1 and -2 was investigated in isolated rat hepatocytes and rat liver microsomes. The toxicity of the compounds was assessed in a murine macrophage cell line by determining cell viability and reduced glutathione (GSH) content. The metabolism and toxicity of flurazepam was assessed for comparison. BNZ-1 and BNZ-2 underwent similar metabolic transformations by the liver systems, forming N-demethylated and hydroxylated metabolites, with subsequent O-glucuronidation. Flurazepam and both analogue compounds depleted macrophage GSH levels without affecting cell viability at the concentrations used (up to 100 microM), but only flurazepam inhibited glutathione reductase activity, indicating that it is acting by a different mechanism. The exact mechanism responsible for GSH depletion is unknown at present. Further experiments are needed to fully understand the effects of BNZs on the parasite GSH analogue, trypanothione, which may be a direct or indirect target for these agents. Pharmacokinetic evaluation of these compounds is required to further progress their development as potential new treatments for leishmaniasis.