Merck
An optical chemical sensor for mercury ion based on 2-mercaptopyrimidine in PVC membrane.
Analytical and bioanalytical chemistry (2008-03-04)
Bahareh Khezri, Mohammad K Amini, Ali R Firooz
RÉSUMÉ
An optical chemical sensor based on 2-mercaptopyrimidine (2-MP) in plasticized poly(vinyl chloride) (PVC) membrane incorporating (N,N-diethyl-5-(octadecanoylimino)-5H benzo[a]phenoxazine-9-amine (ETH 5294) and sodium tetraphenyl borate (NaTPB) for batch and flow-through determination of mercury ion is described. The response of the sensor is based on selective complexation of Hg(2+) with 2-MP in the membrane phase, resulting in an ion exchange process between H(+) in the membrane and Hg(2+) in the sample solution. The influences of several experimental parameters, such as membrane composition, pH, and type and concentration of the regenerating reagent, were investigated. The sensor has a response range of 2.0 x 10(-9) to 2.0 x 10(-5) mol L(-1) Hg(2+) with a detection limit of 4.0 x 10(-10) mol L(-1) and a response time of < or = 45 s at optimum pH of 6.5 with high measurement repeatability and sensor-to-sensor reproducibility. It shows high selectivity for Hg(2+) over several transition metal ions, including Ag(+), Cd(2+), Co(2+), Cr(3+), Cu(2+), Fe(3+), Mn(2+), Ni(2+), and common alkali and alkaline earth ions such as Na(+), K(+), Mg(2+), Ca(2+), and Pb(2+). The sensor membrane can be easily regenerated with dilute acid solutions. The sensor has been used for the determination of mercury ion concentration in water samples.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-Mercaptopyrimidine, 98%
Le tarif et la disponibilité ne sont pas disponibles actuellement.