Merck
Addressing ceiling effects in health status measures: a comparison of techniques applied to measures for people with HIV disease.
Health services research (2008-01-24)
I-Chan Huang, Constantine Frangakis, Mark J Atkinson, Richard J Willke, Walter L Leite, W Bruce Vogel, Albert W Wu
RÉSUMÉ
To compare different approaches to address ceiling effects when predicting EQ-5D index scores from the 10 subscales of the MOS-HIV Health Survey. Data were collected from an HIV treatment trial. Statistical methods included ordinary least squares (OLS) regression, the censored least absolute deviations (CLAD) approach, a standard two-part model (TPM), a TPM with a log-transformed EQ-5D index, and a latent class model (LCM). Predictive accuracy was evaluated using percentage of absolute error (R(1)) and squared error (R(2)) predicted by statistical methods. A TPM with a log-transformed EQ-5D index performed best on R(1); a LCM performed best on R(2). In contrast, the CLAD was worst. Performance of the OLS and a standard TPM were intermediate. Values for R(1) ranged from 0.33 (CLAD) to 0.42 (TPM-L); R(2) ranged from 0.37 (CLAD) to 0.53 (LCM). The LCM and TPM with a log-transformed dependent variable are superior to other approaches in handling data with ceiling effects.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Delavirdine mesylate, ≥98% (HPLC)