Merck
  • 1-phenylethyl isocyanate is a powerful reagent for the chiral analysis of secondary alcohols and hydroxy fatty acids with remote stereogenic centres.
1-phenylethyl isocyanate is a powerful reagent for the chiral analysis of secondary alcohols and hydroxy fatty acids with remote stereogenic centres.
Journal of chromatography. A (2007-08-19)
Andreas Habel, Dieter Spiteller, Wilhelm Boland
RÉSUMÉ
1-phenylethyl isocyanate (1-PEIC), a chiral derivatisation reagent for the resolution of secondary alcohols is a powerful tool to determine the configuration and enantiomeric excess of medium- to long-chain secondary alcohols by capillary gas chromatography. The separation of 1-phenylethylcarbamates (1-PECs) of secondary alcohols was systematically evaluated depending on the position of the stereogenic centre in the molecule, namely in alkanols (C(15)-C(18)), alkenols (C(15)-C(18)) and hydroxy fatty acids (C(14)-C(18)). The successful separation of the diastereomeric carbamates of (+/-)-heptadecan-7-ol or (+/-)-12-hydroxyoctadecanoic acid methyl ester by gas liquid chromatography demonstrates the unique separation power for 1-PECs for analytes with remote stereogenic centres. Saturated derivatives showed consistently higher resolution factors than the corresponding unsaturated derivatives.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Phenethyl isocyanate, 98%