Merck
Preparation of floating microspheres for fish farming.
International journal of pharmaceutics (2007-05-09)
Pushp R Nepal, Myung-Kwan Chun, Hoo-Kyun Choi
RÉSUMÉ
The aim of this study was to develop floating microspheres with practical applications to fish farming. Each microsphere with a central hollow cavity was prepared using a solvent diffusion and evaporation method with Eudragit E100. Various manufacturing parameters were investigated by single factor method. The macrolide antibiotic josamycin was selected as a model drug. The loading efficiency of the drug in the microspheres was 64.7%. In the release study, virtually none of the drug was released into the fresh water whereas the entire drug was released from the josamycin-loaded microspheres into the simulated gastric fluid of rainbow trout (pH 2.7). The buoyancy was excellent with approximately 90% of the microspheres still floating after 24h.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Josamycin, ≥90% (HPLC)
Josamycin, European Pharmacopoeia (EP) Reference Standard