Merck
Identification of anhydroecgonine ethyl ester in the urine of a drug overdose victim.
Journal of forensic sciences (2005-12-31)
Alan L Myers, Heather E Williams, James C Kraner, Patrick S Callery
RÉSUMÉ
Toxicological evaluation of postmortem urine collected from a 41-year-old deceased white male detected anhydroecgonine ethyl ester (ethylecgonidine, AEEE), a transesterification product of smoked cocaine co-abused with ethanol. A solid phase extraction (SPE) method was used to extract cocaine, AEEE, and related metabolites from urine. SPE on a 1 mL urine sample from the decedent followed by GC-MS detected AEEE. Other metabolites identified by GC-MS included cocaine, cocaethylene, and anhydroecgonine methyl ester (AEME). To determine whether some or all of the AEEE was artifactually produced in the heated GC injector port, an alternative LC-MS method was developed. LC/MS following SPE found at least 50 ng/mL of AEEE in the extract. The mass fragmentation (MS/MS and MS3) of AEEE detected in the urine was compared to spectra of authentic, synthesized compound. AEEE is a potential additional forensic marker for the co-abuse of smoked cocaine and ethanol.