- Dalsympathetin--a new isoflavone gentiobioside from Dalbergia sympathetica (Dennst.).
Dalsympathetin--a new isoflavone gentiobioside from Dalbergia sympathetica (Dennst.).
Natural product research (2005-12-02)
N S Nagarajan, M G Sethuraman, C N Manoj, R Priya Rao
PMID16319013
RÉSUMÉ
Two isoflavonoids isolated from Dalbergia sympathetica were identified as 5,4'-dihydroxy-7-methoxyisoflavone (1) (Prunetin) and Prunetin-4'-O-beta-D-gentiobioside (2) (Dalsympathetin). The natural occurrence of Dalsympathetin is reported for the first time. The position of glycosylation in Dalsympathetin at 4'-position has been confirmed by 2D-NMR spectral studies.
MATÉRIAUX