Merck
New (1-deaza)purine derivatives via efficient C-2 nitration of the (1-deaza)purine ring.
Nucleosides, nucleotides & nucleic acids (2004-12-02)
M J Wanner, B Rodenko, M Koch, G J Koomen
RÉSUMÉ
Nitration of substituted (1-deaza)purines using a mixture of tetrabutylammonium nitrate (TBAN) and trifluoracetic acid anhydride (TFAA) was applied to prepare nitrosubstituted (1-deaza)purines at low temperature. The nitro group influences the system twofold: 1) it activates other substituents towards nucleophilic aromatic substitution and 2) it can be substituted itself leading to a variety of di-substituted (1-deaza)purines, also via solid phase syntheses. Several of the molecules obtained were studied for their antiprotozoal activity and for interactions with the different human adenosine receptors.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Azabenzimidazole, 99%