Merck
  • Regulation of kinase cascades and transcription factors by a poly(ADP-ribose) polymerase-1 inhibitor, 4-hydroxyquinazoline, in lipopolysaccharide-induced inflammation in mice.
Regulation of kinase cascades and transcription factors by a poly(ADP-ribose) polymerase-1 inhibitor, 4-hydroxyquinazoline, in lipopolysaccharide-induced inflammation in mice.
The Journal of pharmacology and experimental therapeutics (2004-03-05)
Balazs Veres, Balazs Radnai, Ferenc Gallyas, Gabor Varbiro, Zoltan Berente, Erzsebet Osz, Balazs Sumegi
RÉSUMÉ
Activation of the nuclear enzyme poly(ADP-ribose) polymerase (PARP) is involved in numerous pathophysiological conditions. Because PARP-1 knockout mice are resistant to endotoxin-induced shock and inhibitors of the enzyme were reported to have similar beneficial properties, we investigated the effect of 4-hydroxyquinazoline (4-HQN), a potent PARP-1 inhibitor, on the modulation of kinase cascades and the regulation of transcription factors in a rodent septic shock model. T2-weighted magnetic resonance imaging showed the pattern of anatomical localization of the inflammatory response in bacterial lipopolysaccharide (LPS)-treated mice and the anti-inflammatory effect of the PARP-1 inhibitor. We have found that 4-HQN activated the phosphatidylinositol 3 (PI3)-kinase/Akt pathway in lung, liver, and spleen, and down-regulated two elements of the MAP kinase system. Namely, it dramatically attenuated the activation of the LPS-induced extracellular signal-regulated kinase (ERK)1/2 and p38 mitogen-activated protein (MAP) kinase in a tissue-specific manner. Furthermore, phosphorylation of p90RSK, a downstream target of ERK1/2, showed a similar pattern of down-regulation as did the phosphorylation of ERK1/2 and p38 after LPS and 4-HQN treatment. As a consequence of the aforementioned effects on the kinase pathways, 4-HQN decreased the activation of transcription factor nuclear factor-kappaB (NF-kappaB) and activator protein 1 (AP-1) in LPS-induced endotoxic shock. Our results provide evidence for the first time that the beneficial effects of PARP inhibition in endotoxic shock, such as attenuation of NF-kappaB- and AP-1 transcription factor activation, are mediated, at least partially, through the regulation of the PI3-kinase/Akt pathway and MAP kinase cascades.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
4-Hydroxyquinazoline, 98%
Le tarif et la disponibilité ne sont pas disponibles actuellement.