Merck
The first occurrence in nature of two compounds from hops.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2003-10-28)
Yan Qu, Zhi-Hui Ding, Ji-Kai Liu
RÉSUMÉ
Two compounds, (p-methoxyphenyl) diphenylmethanol (1) and tribenzylamine (2), were isolated from Humulus lupulus. Their structures were established on the basis of spectral evidence (MS, IR, NMR, HMBC, HMQC, 1H-1H COSY experiments). Compounds 1 and 2 were found as natural products at the first time.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Tribenzylamine, ≥99.0% (NT)