Merck
More chemistry of the thaxtomin phytotoxins.
Phytochemistry (2003-10-22)
Russell R King, C Harold Lawrence, Jean Embleton, Larry A Calhoun
RÉSUMÉ
Chemical and biochemical studies indicated the possible involvement of N-acetyltryptophan and 4-nitrotryptophan as intermediates in biosynthesis of the thaxtomin phytotoxins. A search for other potential pathways indirectly resulted in the identification of three unusual thaxtomin analogues derived from the o-thaxtomin A isomer. Investigations to resolve the identity of a previously described thaxtomin A di-glucoside were not supportive of the proposed structure.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
N-Acetyl-L-tryptophan
Supelco
N-Acetyl-L-tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material