Merck
Separation and determination of aminohalogenbenzophenones by high-performance liquid chromatography with electrochemical detection.
Journal of chromatography (1992-07-01)
F J Rodríguez, R M Jiménez, R M Alonso
RÉSUMÉ
A high-performance liquid chromatographic method with electrochemical detection has been developed for the determination of three aminohalogenbenzophenones: 2-amino-2',5-dichlorobenzophenone, 2-amino-5-chlorobenzophenone and 2-amino-5-bromo-2'-fluorobenzophenone, metabolites of benzodiazepinooxazoles and other psychotropic drugs. A mobile phase of methanol-water (65:35), containing 5 mM KH2PO4 appeared to be the optimal when a 4-microns, 60-A Nova-Pak C18 column and a flow-rate of 0.75 ml/min (130 bar) were used. The temperature was optimized at 30 degrees C. The amperometric detector, equipped with glassy carbon electrode, was operated at 1.3 V versus Ag/AgCl in the DC mode. The method was applied to the determination of these compounds at two concentration levels: ppm and ppb (ng/cm3) using 2-amino-5-chlorobenzophenone as internal standard. The limit of determination was 750 pg/ml of biological fluid for each compound, and recoveries greater than 97% were obtained for spiked samples of urine and serum, using C18 Sep-Pak cartridges in the sample clean-up procedure.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-Amino-5-chlorobenzophenone, 98%