Merck
  • 4-Dimethylaminobenzylamine as a sensitive chemiluminescence derivatization reagent for 5-hydroxyindoles and its application to their quantification in human platelet-poor plasma.
4-Dimethylaminobenzylamine as a sensitive chemiluminescence derivatization reagent for 5-hydroxyindoles and its application to their quantification in human platelet-poor plasma.
Journal of chromatography. B, Biomedical sciences and applications (2000-03-16)
J Ishida, M Takada, N Hitoshi, R Iizuka, M Yamaguchi
RÉSUMÉ
A selective and sensitive high-performance liquid chromatographic method with chemiluminescence detection for the determination of 5-hydroxyindoles is described, based on the reaction of 5-hydroxyindoles with 4-dimethylaminobenzylamine. Serotonin, 5-hydroxyindole-3-acetic acid, 5-hydroxytryptophol, 5-hydroxyindole-3-acetamide and N-acetyl-5-hydroxytryptamine were used as model compounds to optimize the derivatization and chemiluminescent reaction. The reagent reacts with 5-hydroxyindoles in slightly alkaline media in the presence of potassium hexacyanoferrate(III) to give the corresponding derivatives, which can be separated on a reversed-phase column, Wakosil-II 5C18RS, with aqueous acetonitrile as an eluent. The derivatives were detected by peroxyoxalate chemiluminescence detection. The detection limits are in the range of 0.5-1.2 fmol per 100-microl injection. The method was applied to the simultaneous determination of serotonin and 5-hydroxyindole-3-acetic acid in human platelet-poor plasma.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-(Dimethylamino)benzylamine dihydrochloride, 95%