Merck
Review and comparison of advanced-generation macrolides clarithromycin and dirithromycin.
Pharmacotherapy (1999-04-22)
S A McConnell, G W Amsden
RÉSUMÉ
We reviewed English-language clinical studies, abstracts, and review articles identified from MEDLINE searches from January 1966-August 1998, and bibliographies of identified articles to compare advanced-generation macrolides dirithromycin and clarithromycin and their use for respiratory tract infections. Both agents have superior adverse effect profiles compared with erythromycin, the original macrolide. Both have broad antibacterial coverage, but clarithromycin usually has a lower MIC90 to susceptible organisms than dirithromycin; for most isolates this difference is not clinically significant. Clarithromycin has better in vitro coverage of Haemophilus influenzae, but this activity varies with formation of its bioactive metabolite, 14-hydroxyclarithromycin. Neither agent is ideal for H. influenzae eradication. The agents differ markedly in terms of pharmacokinetics, pharmacodynamics, metabolism, and cost, and thus with respect to drug interaction profiles and dosages. Dirithromycin's drug interaction profile is markedly better than clarithromycin's. Clarithromycin is dosed twice/day; dirithromycin's pharmacokinetics allow once/day dosing. Dirithromycin is less expensive with regard to both cost/day and cost/treatment regimen. Clarithromycin has been studied and approved for administration to children. In adults with respiratory tract infections who are receiving drugs that would interact with clarithromycin, and in those with renal dysfunction with or without coexisting hepatic dysfunction, dirithromycin appears to be superior in terms of safety and equivalent to clarithromycin in terms of efficacy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Dirithromycin, European Pharmacopoeia (EP) Reference Standard