Merck
Overcoming temozolomide resistance in glioblastoma via dual inhibition of NAD+ biosynthesis and base excision repair.
Cancer research (2011-03-17)
Eva M Goellner, Bradford Grimme, Ashley R Brown, Ying-Chih Lin, Xiao-Hong Wang, Kelsey F Sugrue, Leah Mitchell, Ram N Trivedi, Jiang-bo Tang, Robert W Sobol
RÉSUMÉ
Glioblastoma multiforme (GBM) is a devastating brain tumor with poor prognosis and low median survival time. Standard treatment includes radiation and chemotherapy with the DNA alkylating agent temozolomide (TMZ). However, a large percentage of tumors are resistant to the cytotoxic effects of the TMZ-induced DNA lesion O(6)-methylguanine due to elevated expression of the repair protein O(6)-methylguanine-DNA methyltransferase (MGMT) or a defect in the mismatch repair (MMR) pathway. Although a majority of the TMZ-induced lesions (N7-methylguanine and N3-methyladenine) are base excision repair (BER) substrates, these DNA lesions are also readily repaired. However, blocking BER can enhance response to TMZ and therefore the BER pathway has emerged as an attractive target for reversing TMZ resistance. Our lab has recently reported that inhibition of BER leads to the accumulation of repair intermediates that induce energy depletion-mediated cell death via hyperactivation of poly(ADP-ribose) polymerase. On the basis of our observation that TMZ-induced cell death via BER inhibition is dependent on the availability of nicotinamide adenine dinucleotide (NAD(+)), we have hypothesized that combined BER and NAD(+) biosynthesis inhibition will increase TMZ efficacy in glioblastoma cell lines greater than BER inhibition alone. Importantly, we find that the combination of BER and NAD(+) biosynthesis inhibition significantly sensitizes glioma cells with elevated expression of MGMT and those deficient in MMR, two genotypes normally associated with TMZ resistance. Dual targeting of these two interacting pathways (DNA repair and NAD(+) biosynthesis) may prove to be an effective treatment combination for patients with resistant and recurrent GBM.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Methoxyamine hydrochloride, 98%
Référence
Conditionnement
Disponibilité
Prix
Quantité