Merck
  • Artificial miniaturization causes eggs laid by crowd-reared (gregarious) desert locusts to produce green (solitarious) offspring in the desert locust, Schistocerca gregaria.
Artificial miniaturization causes eggs laid by crowd-reared (gregarious) desert locusts to produce green (solitarious) offspring in the desert locust, Schistocerca gregaria.
Journal of insect physiology (2009-06-10)
Koutaro Maeno, Seiji Tanaka
RÉSUMÉ
The mechanism underlying the phase-dependent polyphenism in hatchling body coloration was studied by testing for a possible causal relationship with egg size in the desert locust, Schistocerca gregaria. Crowd-reared (gregarious) females typically produce large, black offspring, whereas females reared in isolation (solitarious) deposit small, green offspring. We first tested for possible genetic differences in the role of egg foam by washing or separating eggs from two strains of locust. No solitarizing effect was found in either of the strains tested, supporting a previous finding, using another laboratory strain, to show that the hatchling body coloration and size are pre-determined in the ovary of the mother and no egg foam factor is involved in the control of the hatchling body coloration. Topical application of fenoxycarb, a juvenile hormone analog (JHA), and implantation of extra corpora allata (CA), taken from Locusta migratoria, caused gregarious female adults of S. gregaria to produce small eggs. Some eggs laid by CA-implanted females produced green hatchlings. All large eggs chosen among those deposited by gregarious females produced black hatchlings. When eggs were either kept on dry filter paper at nearly saturated relative humidity during embryogenesis or pricked with a needle so that some egg yolk was squeezed out, some produced small, green hatchlings. These results suggested that the amount of egg yolk or the availability of yolk material may determine the body coloration of hatchlings.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Supelco
Fenoxycarb, PESTANAL®, analytical standard