Merck
Triacontanol negatively modulates the jasmonic acid-stimulated proteinase inhibitors in tomato (Lycopersicon esculentum).
Journal of plant physiology (2004-05-07)
Krishnamurthy Ramanarayan, Gangadharamurthy Sivakumar Swamy
RÉSUMÉ
Triacontanol (TRIA), a long chain aliphatic alcohol (C30H61OH) reverses the effect of jasmonic acid (JA) in inducing proteinase inhibitors (PIs) in tomato leaves. Porcine pancreas trypsin and Spodoptera litura gut proteinases were inhibited in the presence of leaf proteins treated with JA, and TRIA partially reverses this effect. Spodoptera litura larvae fed with tomato leaves treated with JA were reduced in body weight and TRIA is able to partially reverse this JA-induced effect. These results reflect the partial reversal effect of TRIA in down regulating the JA-induced production of proteinase inhibitors.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Triacontanol, ≥98% (capillary GC)