Merck
  • Synthesis of novel succinamide derivatives having the 5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one skeleton as potent and selective M2 muscarinic receptor antagonists. I.
Synthesis of novel succinamide derivatives having the 5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one skeleton as potent and selective M2 muscarinic receptor antagonists. I.
Chemical & pharmaceutical bulletin (1997-06-01)
T Watanabe, I Kinoyama, A Kakefuda, T Okazaki, K Takizawa, S Hirano, H Shibata, I Yanagisawa
RÉSUMÉ
A series of 5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one derivatives containing the succinamide skeleton has been synthesized and evaluated for M1, M2 and M3 muscarinic receptor binding affinities (in vitro) and M2 and M3 muscarinic receptor antagonistic activities (in vivo). Some of them showed higher and more selective binding affinities for M(2) muscarinic receptors than that of AF-DX 116. Among them, 11-[3-[N-[2-(N-benzyl-N- methylamino)ethyl]-N-ethylcarbamoyl]propionyl]-5,11-dihydro-6H-pyr ido [2,3-b][1,4]benzodiazepin-6-one (68) was found to be the most potent and selective M2 muscarinic receptor antagonist in vitro. This compound also strongly inhibited the oxotremorine-induced bradycardia after intravenous administration and showed 130-fold selectivity for M2 muscarinic receptors over M3 muscarinic receptors in vivo.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Succinamide, 98%