Merck
Effects of inhibitors on monoamine oxidase activity in perch (Perca flavescens) brain in vitro.
Comparative biochemistry and physiology. C: Comparative pharmacology (1982-01-01)
T R Hall, J M Olcese, H R Figueroa, V L deVlaming
RÉSUMÉ
1. Perch brain homogenates were incubated in vitro and monoamine oxidase (MAO) activity was determined fluorometrically, using a kynuramine substrate. 2. Clorgyline, harmaline and deprenyl inhibited MAO activity in a concentration-related manner, with single sigmoid inhibition curves, and the type A inhibitors harmaline and clorgyline were more effective than the type B inhibitor deprenyl. 3. Two types of inhibition were recognized in vitro; a fast-onsetting inhibition, similar to that produced by a reversible inhibitor, and a slow-onsetting inhibition, which is time- and concentration-dependent and presumably represents inactivation of the enzyme.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
4-Quinolinol, 98%