Merck
  • Extraction of organic acids by ion-pair formation with tri-n-octylamine. Part 6. Determination of sorbic acid, benzoic acid, and saccharin in yogurt.
Extraction of organic acids by ion-pair formation with tri-n-octylamine. Part 6. Determination of sorbic acid, benzoic acid, and saccharin in yogurt.
Journal - Association of Official Analytical Chemists (1985-01-01)
M L Puttemans, C Branders, L Dryon, D L Massart
RÉSUMÉ
The sorbate content of commercial yogurt samples is determined by reverse phase liquid chromatography following ion-pair extraction with tri-n-octylamine. Mean recoveries (70-88%), precision (1.1-3.3% RSD), and detection limit of the method are presented for sorbic acid, benzoic acid, and saccharin.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Trioctylamine, 98%