Merck
Algal growth and community structure in a mixed-culture system using coal seam gas water as the water source.
Environmental technology (2013-07-11)
Jessica J Buchanan, Frances R Slater, Xue Bai, Steven Pratt
RÉSUMÉ
Coal seam gas (CSG) is being touted as a transition fuel as the world moves towards low-carbon economies. However, the development of CSG reserves will generate enormous volumes of saline water. In this work, we investigate the potential of using this saline water to support mass algae production. Water and brine from a CSG water treatment facility (1.6 and 11.6 g total dissolved solids per litre (TDS L(-1)) respectively) were inoculated with algal biomass from freshwater and seawater environments and supplemented with nutrients in open, fed-batch reactors. Significant algal growth was recorded, with maximum specific growth rates in CSG water and CSG brine of 0.20 +/- 0.05 d(-1) and 0.26 +/- 0.04 d(-1) respectively. These maximum specific growth rates were equal to or greater than specific growth rates in deionized water and seawater diluted to the same salinity. However, algal growth lag time in CSG brine was between 7 and 9 times longer than in other waters. Microscopy and terminal-restriction fragment length polymorphism (T-RFLP) were used to monitor community structure in the reactors. The same few algal species dominated all of the reactors, except for the CSG brine reactor at day 15. This result indicates that conditions in CSG brine select for different species of algae compared to seawater of the same salinity and other waters tested. The findings suggest that mass algae production in CSG water is feasible but algae community composition may be a function of CSG water chemistry. This has implications for the downstream use of algae.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Chlorure de sodium, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium solution, 0.9% in water, BioXtra, suitable for cell culture
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Référence
Conditionnement
Disponibilité
Prix
Quantité
SAFC
Chlorure de sodium solution, 5 M
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium solution, BioUltra, for molecular biology, ~5 M in H2O
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium, 99.999% trace metals basis
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium, BioUltra, for molecular biology, ≥99.5% (AT)
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium, BioXtra, ≥99.5% (AT)
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium solution, 5 M
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium, random crystals, optical grade, 99.9% trace metals basis
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium solution, 0.85%
Référence
Conditionnement
Disponibilité
Prix
Quantité
Supelco
Chlorure de sodium, reference material for titrimetry, certified by BAM, >99.5%
Référence
Conditionnement
Disponibilité
Prix
Quantité
Supelco
Chlorure de sodium, Pharmaceutical Secondary Standard; Certified Reference Material
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium, tablet
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chlorure de sodium, tested according to Ph. Eur.
Référence
Conditionnement
Disponibilité
Prix
Quantité