Merck
Anti-inflammatory effect and selectivity profile of AS1940477, a novel and potent p38 mitogen-activated protein kinase inhibitor.
European journal of pharmacology (2012-11-28)
Masaomi Terajima, Tatsuo Inoue, Katsue Magari, Hitoshi Yamazaki, Yasuyuki Higashi, Hidekazu Mizuhara
RÉSUMÉ
Given the key role p38 mitogen-activated protein kinase (MAPK) plays in inflammatory responses through the production of cytokines and inflammatory mediators, its inhibition is considered a promising therapeutic strategy for chronic inflammatory diseases such as rheumatoid arthritis, psoriasis, inflammatory bowel disease, and chronic obstructive pulmonary disease. Here, we evaluated the anti-inflammatory effect and selectivity profile of the novel p38 MAPK inhibitor AS1940477. AS1940477 inhibited the enzymatic activity of recombinant p38α and β isoforms but showed no effect against other 100 protein kinases including p38γ and δ isoforms. We also confirmed the selectivity of AS1940477 in the intracellular signaling pathway. In human peripheral blood mononuclear cells, AS1940477 inhibited lipopolysaccharide (LPS)- or phytohemagglutinin A (PHA)-induced production of proinflammatory cytokines, including TNFα, IL-1β, and IL-6 at low concentrations (LPS/TNFα, IC(50)=0.45n M; PHA/TNFα, IC(50)=0.40 nM). In addition, equivalent concentrations of AS1940477 that inhibited cytokine production also inhibited TNFα- and IL-1 β-induced production of IL-6, PGE(2), and MMP-3 in human synovial stromal cells. AS1940477 was also found to potently inhibit TNF production in whole blood (IC(50)=12 nM) and effectively inhibited TNFα production induced by systemically administered LPS in rats at less than 0.1mg/kg (ED(50)=0.053 mg/kg) with an anti-inflammatory effect lasting for 20h after oral administration. Overall, this study demonstrated that AS1940477 is a novel and potent p38 MAPK inhibitor and may be useful as a promising anti-inflammatory agent for treating inflammatory disorders.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Pyridazine, 98%